Learn More
INTACT GENOMICS INC sgRNA Synthesis Kit for Cas12a Nuclease 10 Reactions (100 µl RXN Volume)
Catalog No. 5000082960
Encompass
IG sgRNA Synthesis Kits are a perfect choice for a variety of sgRNA synthesis needs. Below is a sampling of some of the key benefits. IG sgRNA Synthesis Kit includes everything but your targeting oligo (all buffers, enzymes, scaffolds) User simply needs to provide the oligo for your target—Must be 55 base pairs and generated using the template design in this manual. Rapid Workflow (Less than 1 hour) Scalable Yield (just 1 reaction for all the sgRNA you need) Customer support – our team is available to aid with your success.IG sgRNA Synthesis Kits contains the items below. Store all components at -20°C. IG sgRNA Enzyme Mix (10X) IG sgRNA Reaction Mix, S. pyogenes (4X) DNase I (RNase-free, 2 Units/uL) IG sgRNA Control Oligo, S. pyogenes (1 uM) Dithiothreitol (DTT, 0.2 M) NTPs mix (25 mM)
Catalog No. 50-000-82960 Supplier INTACT GENOMICS INC Supplier No. 3306
IG sgRNA Synthesis Kits are a perfect choice for a variety of sgRNA synthesis needs. Below is a sampling of some of the key benefits. IG sgRNA Synthesis Kit includes everything but your targeting oligo (all buffers, enzymes, scaffolds) User simply needs to provide the oligo for your target—Must be 55 base pairs and generated using the template design in this manual. Rapid Workflow (Less than 1 hour) Scalable Yield (just 1 reaction for all the sgRNA you need) Customer support – our team is available to aid with your success.IG sgRNA Synthesis Kits contains the items below. Store all components at -20°C. IG sgRNA Enzyme Mix (10X) IG sgRNA Reaction Mix, S. pyogenes (4X) DNase I (RNase-free, 2 Units/uL) IG sgRNA Control Oligo, S. pyogenes (1 uM) Dithiothreitol (DTT, 0.2 M) NTPs mix (25 mM)