Learn More
Aviva Systems Biology Corp SGTA Antibody - C-terminal region (ARP64527_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP64527_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SGTA. It was validated on Western Blot. . Alternate names: SGT, alphaSGT, hSGTApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rat
Catalog No. 50-288-283