Learn More
Selleck Chemical LLC SGX-523 5mg 1022150-57-7
SDP
Supplier:  Selleck Chemical LLC S11125MG
Encompass_Preferred
SGX-523 is a selective Met (c-Met) inhibitor with IC50 of 4 nM, no activity to BRAFV599E, c-Raf, Abl and p38?. Phase 1. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-853-8