Learn More
Selleck Chemical LLC SH-4-54
SDP
Catalog No. 5000004229
Encompass_Preferred
SH-4-54 is a potent STAT inhibitor with KD of 300 nM and 464 nM for STAT3 and STAT5 respectively
Catalog No. 50-000-04229 Supplier Selleck Chemical LLC Supplier No. S7337100MG
SH-4-54 is a potent STAT inhibitor with KD of 300 nM and 464 nM for STAT3 and STAT5 respectively