Learn More
Selleck Chemical LLC SH-4-54 5mg 1456632-40-8
SDP
Supplier:  Selleck Chemical LLC S73375MG
Encompass_Preferred
SH-4-54 is a potent STAT inhibitor with KD of 300 nM and 464 nM for STAT3 and STAT5, respectively. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC1072019