Learn More
Innovative Research Inc Sh FX Ply Frctn FITC lbl 100ug
Catalog No. 502743961
Encompass_Preferred
Sheep Anti Mouse Factor X Polyclonal Fractionated FITC Labeled 100ug - Sheep Anti Mouse Factor X Polyclonal Fractionated FITC Labeled from Innovative Research is a polyclonal antibody in a Frozen liquid format. This antibody has been purified using protein G affinity chromatography followed by fluorescein isothiocyanate conjugation. Fractionated preparations may be especially useful in research where high affinity is necessary, especially in cases where the target antigen is less abundant. IgG Fractionated antiserum can also contain very high affinity antibodies that are sometimes missing from affinity purified preparations (since they can bind so tightly to the affinity matrix that they are not always eluted).
Catalog No. 50-274-3961 Supplier Innovative Research Inc Supplier No. ISHAMSFXGFFITC100UG
Sheep Anti Mouse Factor X Polyclonal Fractionated FITC Labeled 100ug - Sheep Anti Mouse Factor X Polyclonal Fractionated FITC Labeled from Innovative Research is a polyclonal antibody in a Frozen liquid format. This antibody has been purified using protein G affinity chromatography followed by fluorescein isothiocyanate conjugation. Fractionated preparations may be especially useful in research where high affinity is necessary, especially in cases where the target antigen is less abundant. IgG Fractionated antiserum can also contain very high affinity antibodies that are sometimes missing from affinity purified preparations (since they can bind so tightly to the affinity matrix that they are not always eluted).