Learn More
Innovative Research Inc Sh tPA Ply Aff Pur Btn lbl
Catalog No. 502743958
Encompass_Preferred
Sheep Anti Mouse/Rat tPA Polyclonal Affinity Purified Biotin Labeled 100ug - Sheep Anti Mouse/Rat tPA Polyclonal Affinity Purified Biotin Labeled from Innovative Research is a polyclonal antibody in a Frozen liquid format buffered in 0.05M Sodium Phosphate; 0.1M NaCl; 1mM EDTA; pH 6.6. The affinity isolated antigen-specific antibody was purified from whole antiserum by immunospecific purification using antigen-bound agarose. This process removes the host serum proteins (including immunoglobulins) that do not specifically bind to the target antigen. This antibody preparation has the lowest amount of non-specific binding and therefore the highest titer. This product can be used in analytic tools such as ELISA kits and Western blot.
Catalog No. 50-274-3958 Supplier Innovative Research Inc Supplier No. ISHAMSTPAAPBL100UG
Sheep Anti Mouse/Rat tPA Polyclonal Affinity Purified Biotin Labeled 100ug - Sheep Anti Mouse/Rat tPA Polyclonal Affinity Purified Biotin Labeled from Innovative Research is a polyclonal antibody in a Frozen liquid format buffered in 0.05M Sodium Phosphate; 0.1M NaCl; 1mM EDTA; pH 6.6. The affinity isolated antigen-specific antibody was purified from whole antiserum by immunospecific purification using antigen-bound agarose. This process removes the host serum proteins (including immunoglobulins) that do not specifically bind to the target antigen. This antibody preparation has the lowest amount of non-specific binding and therefore the highest titer. This product can be used in analytic tools such as ELISA kits and Western blot.