Learn More
Cayman Chemical (S)-H-1152 (hydrochloride), Purity: ≥95%, CAS: 451462-58-1, 1 mg
SDP
Catalog No. 502891337
Encompass_Preferred
A potent, specific, ATP-competitive, and cell permeable inhibitor of ROCK (Ki = 1.6 nM); more potent inhibitor of ROCK than either Y-27632 (Ki = 140 nM) or HA-1077 (Ki = 330 nM); poorly inhibits PKA, PKC, and MLCK
Catalog No. 50-289-1337 Supplier Cayman Chemical Supplier No. 100076531 MG
A potent, specific, ATP-competitive, and cell permeable inhibitor of ROCK (Ki = 1.6 nM); more potent inhibitor of ROCK than either Y-27632 (Ki = 140 nM) or HA-1077 (Ki = 330 nM); poorly inhibits PKA, PKC, and MLCK