Learn More
Aviva Systems Biology Corp SH2B1 Antibody - middle region (ARP55271_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55271_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SH2B1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human SH2B1. Alternate names: DKFZp547G1110, FLJ30542, SH2-B, SH2B, PSMApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-61453