Learn More
Aviva Systems Biology Corp SH2D1A Antibody - C-terminal region (ARP60359_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60359_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SH2D1A. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human SH2D1A. Alternate names: DSHP, EBVS, FLJ18687, FLJ92177, IMD5, LYP, MTCP1, SAP, XLP, XLPD, SAP/SH2D1AApplicationWB. Species reactivity: Cow, Horse, Human, Pig, Rat
Catalog No. 50-287-019