Learn More
Aviva Systems Biology Corp SH3BP2 Antibody - middle region (ARP56546_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56546_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SH3BP2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human SH3BP2. Alternate names: 3BP2, CRBM, CRPM, FLJ42079, RES4-23, 3BP-2ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-61898