Learn More
Aviva Systems Biology Corp SH3BP4 Antibody - N-terminal region (ARP55075_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55075_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SH3BP4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human SH3BP4. Alternate names: BOG25, TTPApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-62978