Learn More
Aviva Systems Biology Corp SH3BP5 Antibody - C-terminal region (ARP60361_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60361_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SH3BP5. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human SH3BP5. Alternate names: SABApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-287-020