Learn More
Aviva Systems Biology Corp SH3GL1 Antibody - N-terminal region (ARP56547_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56547_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SH3GL1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human SH3GL1. Alternate names: CNSA1, EEN, MGC111371, SH3D2B, SH3P8ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-61899