Learn More
Aviva Systems Biology Corp Sh3gl3 Antibody - middle region (ARP56551_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56551_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Sh3gl3. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: EEN-B2, MGC118269, SH3P13, Sh3d2c, Sh3d2c2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-286-175