Learn More
Aviva Systems Biology Corp SH3KBP1 Antibody - N-terminal region (ARP54453_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP54453_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SH3KBP1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human SH3KBP1. Alternate names: CIN85, GIG10, MIG18, HSB1, HSB-1, CD2BP3ApplicationWB, IHC. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-285-927