Learn More
Enzo Life Sciences SH5 (0.5mg)
SDP
Supplier:  Enzo Life Sciences ALX270349MC05
Encompass_Preferred
Inhibitor of Akt (PKB) activation without affecting activation of the upstream kinase PDK-1, or other kinases downstream of Ras such as MAPK. Induces apoptosis and kills a variety of cancer cell lines containing high levels of active Akt and depending upon Akt for survival. Alternative name: D-3-Deoxy-2-O-methyl-myo-inositol 1-[(R)-2-methoxy-3-(octadecyloxy)propyl hydrogen phosphate]. Formula: C29H59O10P. MW: 598.7. Purity: ≥98% (TLC). Solubility: Soluble in DMSO (10mg/ml) or methanol; slightly soluble in water. Long Term Storage: +4°C.
Catalog No. 50-200-8498