Learn More
Sony Biotechnology SH800 Cell Sorter with 488nm/405nm/561nm/638nm Laser and 96 well plate deposition system
Catalog No. NC3885106
Encompass_Preferred
The benchtop SH800S Cell Sorter enables precise sorting across a wide range of cell sizes using 70 μm, 100 μm, and 130 μm microfluidic chips for sorting into 2-way tubes and multi-well plates (96 and 384 well). Its innovative chip-based design integrates advanced fluidics control and automation for setup, acquisition, sorting, and analysis making the process more accurate, consistent, and user-friendly. Intuitive software and guided workflows simplify startup, multicolor compensation, sorting, and system maintenance. Patented CoreFinder technology automates key setup steps of system and stream setup, sorting and monitoring streamlining operation. An optional class II biosafety cabinet custom designed for the SH800S provides product, personnel and environment protection while sorting biohazardous samples.
Sony SH800S Cell Sorter Brochure
Catalog No. NC3885106 Supplier Sony Biotechnology Supplier No. LESH800SFP
This product is not available for purchase online. Please contact your sales representative for a quote or further ordering instructions.
The benchtop SH800S Cell Sorter enables precise sorting across a wide range of cell sizes using 70 μm, 100 μm, and 130 μm microfluidic chips for sorting into 2-way tubes and multi-well plates (96 and 384 well). Its innovative chip-based design integrates advanced fluidics control and automation for setup, acquisition, sorting, and analysis making the process more accurate, consistent, and user-friendly. Intuitive software and guided workflows simplify startup, multicolor compensation, sorting, and system maintenance. Patented CoreFinder technology automates key setup steps of system and stream setup, sorting and monitoring streamlining operation. An optional class II biosafety cabinet custom designed for the SH800S provides product, personnel and environment protection while sorting biohazardous samples.
Sony SH800S Cell Sorter Brochure