Learn More
MSC SHADE 8-12 3M
Catalog No. 5000684925
Encompass
This polycarbonate filter lens is designed for welding applications, providing variable shade protection from 8 to 12. It features a green lens color for optimal visibility.
  • Variable lens shade from 8 to 12
  • Green lens color for enhanced clarity
  • Constructed from polycarbonate for durability
  • Designed for inside mounting
  • Window height of 1.7 in
  • Window width of 3.7 in
  • Window thickness of 0.0630 in
Catalog No. 50-006-84925 Supplier MSC Supplier No. 59055996
This polycarbonate filter lens is designed for welding applications, providing variable shade protection from 8 to 12. It features a green lens color for optimal visibility.
  • Variable lens shade from 8 to 12
  • Green lens color for enhanced clarity
  • Constructed from polycarbonate for durability
  • Designed for inside mounting
  • Window height of 1.7 in
  • Window width of 3.7 in
  • Window thickness of 0.0630 in