Learn More
High-Tech Conversions SHADOW Black Nitrile Exam Glove
Description
- Ambidextrous
- Single Use Only
- Powder-free
- 100 per box (90 per XL and XXL box)
Specifications
Specifications
| Chemotherapeutic Rated | No |
| Chlorinated | No |
| Accelerator Free | No |
| Applications | Industrial |
| AQL | 1.5 |
| Color | Black |
| Disposable | Single-use |
| Exterior Finish | Fully Textured |
| Finger Thickness (English) | 6 mil |
| Finger Thickness (Metric) | 0.15 mm |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.