Learn More
Grainger Shadow Marking Tape Green 12inx12ft Roll
Catalog No. 502749101
Encompass_Preferred
The Brady Shadow Marking Tape is a general purpose marking tape that creates dark, opaque lines on light surfaces for labeling and identification. It has an acrylic adhesive and is made of 6 mil polyester. The tape comes in multiple widths from 1/4 inch to 1 inch and in lengths of 36 yards. It can withstand temperatures up to 200°F. The tape creates neat, uniform lines for marking bins, shelves, floors, and more. It sticks well to clean, dry surfaces like metal, plastic, glass and painted wood.
Catalog No. 50-274-9101 Supplier Grainger Supplier No. 14D468
The Brady Shadow Marking Tape is a general purpose marking tape that creates dark, opaque lines on light surfaces for labeling and identification. It has an acrylic adhesive and is made of 6 mil polyester. The tape comes in multiple widths from 1/4 inch to 1 inch and in lengths of 36 yards. It can withstand temperatures up to 200°F. The tape creates neat, uniform lines for marking bins, shelves, floors, and more. It sticks well to clean, dry surfaces like metal, plastic, glass and painted wood.