Learn More
Grainger Shadow Marking Tape Red 12 inx12 ft Roll
Catalog No. 502752029
Encompass_Preferred
The Brady Shadow Marking Tape is a general purpose marking tape that creates dark, opaque lines on light surfaces for labeling and identification. It has an acrylic adhesive and is made of 6 mil polyester. The tape comes in multiple widths from 1/4 inch to 1 inch and in lengths of 60 yards. It can withstand temperatures up to 200°F. The tape creates neat, uniform lines for marking bins, shelves, floors, and more. It sticks well to clean, dry surfaces like metal, glass, plastic, painted wood, and more.
Catalog No. 50-275-2029 Supplier Grainger Supplier No. 14D473
The Brady Shadow Marking Tape is a general purpose marking tape that creates dark, opaque lines on light surfaces for labeling and identification. It has an acrylic adhesive and is made of 6 mil polyester. The tape comes in multiple widths from 1/4 inch to 1 inch and in lengths of 60 yards. It can withstand temperatures up to 200°F. The tape creates neat, uniform lines for marking bins, shelves, floors, and more. It sticks well to clean, dry surfaces like metal, glass, plastic, painted wood, and more.