Learn More
MSC Whitney Rand Standard Shadow Shelving, 14 Gauge, 5 Shelves, 1 ea
Catalog No. 5000510398
Encompass
This standard shelving unit provides a durable storage solution for industrial and commercial applications. Engineered from high-quality 14-gauge steel, the unit offers robust support for heavy-duty material handling and organizational needs.
  • Constructed from heavy-duty 14-gauge steel
  • Features a 5-shelf configuration
  • Overall dimensions of 48 in. (W) x 24 in. (D) x 84 in. (H)
Catalog No. 50-005-10398 Supplier MSC Supplier No. 89841225
This standard shelving unit provides a durable storage solution for industrial and commercial applications. Engineered from high-quality 14-gauge steel, the unit offers robust support for heavy-duty material handling and organizational needs.
  • Constructed from heavy-duty 14-gauge steel
  • Features a 5-shelf configuration
  • Overall dimensions of 48 in. (W) x 24 in. (D) x 84 in. (H)