Learn More
Pipette Supplies, Inc. Rainin Pipet-Lite / Pipet-Lite XLS Shaft Coupling Nut, Single Channel Pipettes, 2µL-1000µL
SDP
Catalog No. NC3667879
Encompass
Shaft coupling nut for the following pipettes:Pipet-Lite, Single Channel, L, SL, 2μL – 1000µLPipet-Lite XLS, Single Channel, XLS, 2μL – 1000µL
Catalog No. NC3667879 Supplier Pipette Supplies, Inc. Supplier No. RA 17011281
Shaft coupling nut for the following pipettes:Pipet-Lite, Single Channel, L, SL, 2μL – 1000µLPipet-Lite XLS, Single Channel, XLS, 2μL – 1000µL