Learn More
Grainger Shaft Grounding Ring For 1 3/8 in Shaft
Catalog No. 502753084
Encompass_Preferred
The AEGIS Shaft Grounding Ring is a one-piece stainless steel ring that provides a path to ground for induced shaft voltages. It has a split-ring design that allows for easy installation without having to remove the shaft. This grounding ring fits shafts from 1-3/4" to 3-15/16" diameter and has a 15kA short duration current rating. It comes with oxide inhibiting conductive grease for installation on the shaft. Overall, this grounding ring provides effective shaft grounding protection.
Catalog No. 50-275-3084 Supplier Grainger Supplier No. 14R031
The AEGIS Shaft Grounding Ring is a one-piece stainless steel ring that provides a path to ground for induced shaft voltages. It has a split-ring design that allows for easy installation without having to remove the shaft. This grounding ring fits shafts from 1-3/4" to 3-15/16" diameter and has a 15kA short duration current rating. It comes with oxide inhibiting conductive grease for installation on the shaft. Overall, this grounding ring provides effective shaft grounding protection.