Learn More
Grainger Shaft Grounding Ring For 1 5/8 in Shaft
Catalog No. 502753108
Encompass_Preferred
The AEGIS Shaft Grounding Ring is a one-piece stainless steel ring that provides a path to ground for induced shaft voltages. It has a split-ring design that allows installation without removing the shaft. This model has a 3.5" shaft diameter, 0.5" ring width, and includes mounting hardware. It is made in the USA and is designed for industrial applications on motors, pumps, fans, blowers, and other rotating shafts where shaft grounding is needed. The ring contacts the shaft for its entire inner circumference for optimal grounding effectiveness.
Catalog No. 50-275-3108 Supplier Grainger Supplier No. 14R032
The AEGIS Shaft Grounding Ring is a one-piece stainless steel ring that provides a path to ground for induced shaft voltages. It has a split-ring design that allows installation without removing the shaft. This model has a 3.5" shaft diameter, 0.5" ring width, and includes mounting hardware. It is made in the USA and is designed for industrial applications on motors, pumps, fans, blowers, and other rotating shafts where shaft grounding is needed. The ring contacts the shaft for its entire inner circumference for optimal grounding effectiveness.