Learn More
Grainger Shaft Grounding Ring For 1 7/8 in Shaft
Catalog No. 502752625
Encompass_Preferred
The AEGIS Shaft Grounding Ring is a one-piece stainless steel ring that provides a path to ground for induced shaft voltages. It has a split-ring design that allows easy installation without shaft removal. This grounding ring fits shafts from 1-3/4" to 3-15/16" diameter and has a durable construction that provides long service life. It comes with oxide-piercing stainless steel mounting screws for solid electrical contact. The one-piece design eliminates the need for hinges or springs. This is an effective and convenient option for shaft grounding in electric motors, generators, turbines, and other rotating equipment.
Catalog No. 50-275-2625 Supplier Grainger Supplier No. 14R033
The AEGIS Shaft Grounding Ring is a one-piece stainless steel ring that provides a path to ground for induced shaft voltages. It has a split-ring design that allows easy installation without shaft removal. This grounding ring fits shafts from 1-3/4" to 3-15/16" diameter and has a durable construction that provides long service life. It comes with oxide-piercing stainless steel mounting screws for solid electrical contact. The one-piece design eliminates the need for hinges or springs. This is an effective and convenient option for shaft grounding in electric motors, generators, turbines, and other rotating equipment.