Learn More
Grainger Shaft Grounding Ring For 2 1/8 in Shaft
Catalog No. 502752948
Encompass_Preferred
The AEGIS Shaft Grounding Ring is a one-piece stainless steel ring that provides a path to ground for induced shaft voltages. It has a split-ring design that allows for easy installation without having to remove the coupling. This grounding ring fits shafts from 1-3/4" to 7-15/16" in diameter and has a durable construction that provides long service life. It comes with mounting hardware for attaching a grounding cable. The AEGIS grounding ring is made in the USA and is ideal for grounding motor shaft voltages to prevent electrical damage.
Catalog No. 50-275-2948 Supplier Grainger Supplier No. 14R034
The AEGIS Shaft Grounding Ring is a one-piece stainless steel ring that provides a path to ground for induced shaft voltages. It has a split-ring design that allows for easy installation without having to remove the coupling. This grounding ring fits shafts from 1-3/4" to 7-15/16" in diameter and has a durable construction that provides long service life. It comes with mounting hardware for attaching a grounding cable. The AEGIS grounding ring is made in the USA and is ideal for grounding motor shaft voltages to prevent electrical damage.