Learn More
Grainger Shaft Grounding Ring For 2 3/8 in Shaft
Catalog No. 502752267
Encompass_Preferred
The AEGIS Shaft Grounding Ring is a one-piece stainless steel ring that provides a path to ground for induced shaft voltages. It has a split-ring design that allows easy installation without shaft removal. This grounding ring fits shafts from 1-3/4" to 3-15/16" diameter and has a durable construction that provides long service life. It comes with oxide-piercing stainless steel mounting screws for solid electrical contact. The one-piece design eliminates the need for hinges or springs. This shaft grounding ring helps protect bearings from electrical damage.
Catalog No. 50-275-2267 Supplier Grainger Supplier No. 14R035
The AEGIS Shaft Grounding Ring is a one-piece stainless steel ring that provides a path to ground for induced shaft voltages. It has a split-ring design that allows easy installation without shaft removal. This grounding ring fits shafts from 1-3/4" to 3-15/16" diameter and has a durable construction that provides long service life. It comes with oxide-piercing stainless steel mounting screws for solid electrical contact. The one-piece design eliminates the need for hinges or springs. This shaft grounding ring helps protect bearings from electrical damage.