Learn More
Bellco Glass, Inc. Shake Flask W/3 Baff,2000mL 38mm Delong Neck
SDP
Catalog No. 501945517
Encompass_Preferred
Manufactured from Type 1 Class A Borosilicate Glass. Shake flasks are ideal for preliminary fermentation and bacterial growth studies. They are the basic tool for growth studies of aerobic microbial cultures. They have rigidly controlled baffle angles and depths to promote optimal oxygen transfer while reducing vortex formation. Designed for all orbital shakers. Closures sold separately.. MADE IN THE USASpecifications:Type 1 Class A Borosilicate GlassSize: 2LDimensions: 160x295mmClosure Sizes: 38Baffle Dimensions: 10mm DP X 35°Case Quantity: 3
Catalog No. 50-194-5517 Supplier Bellco Glass, Inc. Supplier No. 254002000
Manufactured from Type 1 Class A Borosilicate Glass. Shake flasks are ideal for preliminary fermentation and bacterial growth studies. They are the basic tool for growth studies of aerobic microbial cultures. They have rigidly controlled baffle angles and depths to promote optimal oxygen transfer while reducing vortex formation. Designed for all orbital shakers. Closures sold separately.. MADE IN THE USASpecifications:Type 1 Class A Borosilicate GlassSize: 2LDimensions: 160x295mmClosure Sizes: 38Baffle Dimensions: 10mm DP X 35°Case Quantity: 3