Learn More
Bellco Glass, Inc. Shake Flask W/3 Baff,250mL 38mm Delong Neck
SDP
Catalog No. 501945513
Encompass_Preferred
SHAKE FLASK W/3 BAFFLES. -IDEAL FOR PRELIMINARY FERMENTATION AND BACTERIAL GROWTH STUDIES -DELONG STYLE NECK AND THREE SIDE BAFFLES BOROSILICATE GLASS TYPE 1, CLASS A -CLOSURES SOLD SEPERATELY . MADE IN THE USA BAFFLE DIMENSIONS: 4mm DP X 35° CASE Qty: 12
Catalog No. 50-194-5513 Supplier Bellco Glass, Inc. Supplier No. 254000250
SHAKE FLASK W/3 BAFFLES. -IDEAL FOR PRELIMINARY FERMENTATION AND BACTERIAL GROWTH STUDIES -DELONG STYLE NECK AND THREE SIDE BAFFLES BOROSILICATE GLASS TYPE 1, CLASS A -CLOSURES SOLD SEPERATELY . MADE IN THE USA BAFFLE DIMENSIONS: 4mm DP X 35° CASE Qty: 12