Learn More
Bellco Glass, Inc. Shake Flask W/3 Baff,300mL 38mm Delong Neck
SDP
Catalog No. 501945514
Encompass_Preferred
Manufactured from Type 1 Class A Borosilicate Glass. Shake flasks are ideal for preliminary fermentation and bacterial growth studies. They are the basic tool for growth studies of aerobic microbial cultures. They have rigidly controlled baffle angles and depths to promote optimal oxygen transfer while reducing vortex formation. Designed for all orbital shakers. Closures sold separately.. MADE IN THE USASpecifications:Type 1 Class A Borosilicate GlassSize: 300mlDimensions: 88x170mmClosure Sizes: 38Baffle Dimensions: 4mm DP X 35°Case Quantity: 12
Catalog No. 50-194-5514 Supplier Bellco Glass, Inc. Supplier No. 254000300
Manufactured from Type 1 Class A Borosilicate Glass. Shake flasks are ideal for preliminary fermentation and bacterial growth studies. They are the basic tool for growth studies of aerobic microbial cultures. They have rigidly controlled baffle angles and depths to promote optimal oxygen transfer while reducing vortex formation. Designed for all orbital shakers. Closures sold separately.. MADE IN THE USASpecifications:Type 1 Class A Borosilicate GlassSize: 300mlDimensions: 88x170mmClosure Sizes: 38Baffle Dimensions: 4mm DP X 35°Case Quantity: 12