Learn More
Bellco Glass, Inc. Shake Flask W/3 Baff,50mL 18mm Delong Neck
SDP
Catalog No. 501945511
Encompass_Preferred
Manufactured from Type 1 Class A Borosilicate Glass. Shake flasks are ideal for preliminary fermentation and bacterial growth studies. They are the basic tool for growth studies of aerobic microbial cultures. They have rigidly controlled baffle angles and depths to promote optimal oxygen transfer while reducing vortex formation. Designed for all orbital shakers. Closures sold separately.. MADE IN THE USASpecifications:Type 1 Class A Borosilicate GlassSize: 50mlDimensions: 50x105mmClosure Sizes: 18Baffle Dimensions: 4 X 35mmCase Quantity: 12
Catalog No. 50-194-5511 Supplier Bellco Glass, Inc. Supplier No. 254000050
Manufactured from Type 1 Class A Borosilicate Glass. Shake flasks are ideal for preliminary fermentation and bacterial growth studies. They are the basic tool for growth studies of aerobic microbial cultures. They have rigidly controlled baffle angles and depths to promote optimal oxygen transfer while reducing vortex formation. Designed for all orbital shakers. Closures sold separately.. MADE IN THE USASpecifications:Type 1 Class A Borosilicate GlassSize: 50mlDimensions: 50x105mmClosure Sizes: 18Baffle Dimensions: 4 X 35mmCase Quantity: 12