Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3B Scientific™ Shaken Baby Demonstration Model
SDPSCI_ED

Catalog No. S00526 Shop All 3B Scientific Products
Change view
Click to view available options
Length (English):
19 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (English)
S00526 19 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S00526 Supplier 3B Scientific™ Supplier No. 1017928
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

An infant's vulnerability to shaken baby syndrome (SBS) is powerfully demonstrated with this 3B Scientific model.

  • An infant's vulnerability to shaken baby syndrome (SBS) is powerfully demonstrated with this model.
  • The model's translucent head allows viewers to see how a baby's brain can be harmed.
  • Also emphasizes the importance of supporting an infant's neck.
  • Helps teach prevention of SBS, which can have devastating consequences including permanent brain damage, blindness, and death.
  • Model is 19 in. long.

Specifications

For Use With (Application) Teach Prevention of SBS
Length (English) 19 in.
Weight (English) 3.75 lb.
Product Type Shaken Baby Demonstration
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.