Learn More
Chemglass Life Sciences 3D Shaker / Rocker
SDP
Catalog No. 501311337
Encompass
3D Shaker / Rocker. • Small footprint• 3D motion provides thorough, gentle mixing• Dimpled mat holds tubes firmly in placeEconomical, 3D shaker / rocker gently mixes liquid samples with a smooth motion that combines the actions of an orbital shaker and a rocker.
Catalog No. 50-131-1337 Supplier Chemglass Life Sciences Supplier No. CLS4029100
3D Shaker / Rocker. • Small footprint• 3D motion provides thorough, gentle mixing• Dimpled mat holds tubes firmly in placeEconomical, 3D shaker / rocker gently mixes liquid samples with a smooth motion that combines the actions of an orbital shaker and a rocker.