Learn More
Bellco Glass, Inc. Shaker Base Only, 115V
SDP
Catalog No. 501946129
Encompass_Preferred
The Bellco Benchtop Orbital Shakers are ideal for staining and washingelectrophoresis gels or for any applications requiring mixing or agitationof solutions. They are engineered to operate between the limits of 30 andapproximately 350 revolutions per minute, and to hold their speed at anyselected setting to approximately plus or minus five percent.. MADE IN THE USA--Speed range: 30 - 350 rpm +/- 5%--Circular orbit: 2.5 cm (1" ) orbit--Load: 11 kg (25 lbs) maximum--Weight: 7.6 kg (17 lbs)
Catalog No. 50-194-6129 Supplier Bellco Glass, Inc. Supplier No. 774401000
The Bellco Benchtop Orbital Shakers are ideal for staining and washingelectrophoresis gels or for any applications requiring mixing or agitationof solutions. They are engineered to operate between the limits of 30 andapproximately 350 revolutions per minute, and to hold their speed at anyselected setting to approximately plus or minus five percent.. MADE IN THE USA--Speed range: 30 - 350 rpm +/- 5%--Circular orbit: 2.5 cm (1" ) orbit--Load: 11 kg (25 lbs) maximum--Weight: 7.6 kg (17 lbs)