Learn More
Bellco Glass, Inc. Shaker Base Only, 230V
SDP
Catalog No. 501946132
Encompass_Preferred
The Bellco Benchtop Orbital Shakers are ideal for staining and washingelectrophoresis gels or for any applications requiring mixing or agitationof solutions. They are engineered to operate between the limits of 30 andapproximately 350 revolutions per minute, and to hold their speed at anyselected setting to approximately plus or minus five percent.. MADE IN THE USA--Speed range: 30 - 350 rpm +/- 5%--Circular orbit: 2.5 cm (1" ) orbit--Load: 11 kg (25 lbs) maximum--Weight: 7.6 kg (17 lbs)
Catalog No. 50-194-6132 Supplier Bellco Glass, Inc. Supplier No. 774402000
The Bellco Benchtop Orbital Shakers are ideal for staining and washingelectrophoresis gels or for any applications requiring mixing or agitationof solutions. They are engineered to operate between the limits of 30 andapproximately 350 revolutions per minute, and to hold their speed at anyselected setting to approximately plus or minus five percent.. MADE IN THE USA--Speed range: 30 - 350 rpm +/- 5%--Circular orbit: 2.5 cm (1" ) orbit--Load: 11 kg (25 lbs) maximum--Weight: 7.6 kg (17 lbs)