Learn More
Kuhner Shaker Inc SHAKER CABINET ISF4-X WITH COO
Catalog No. 502570050
Encompass
Shaker Cabinet ISF4-X with cooling..Temperature range: ambient - 15C to 80C..Control of 0.1C, Accuracy +/- 0.3C at 37C..100% stainless steel interior..External Dimensions: H1916 x W1494 x D960mm..Weight: 750kg..FIVE YEAR WARRANTY HARDWARE, 1 YEAR WARRA...
Catalog No. 50-257-0050 Supplier Kuhner Shaker Inc Supplier No. SMX1601
Add to Cart
Add to Cart
Request a Quote
Shaker Cabinet ISF4-X with cooling..Temperature range: ambient - 15C to 80C..Control of 0.1C, Accuracy +/- 0.3C at 37C..100% stainless steel interior..External Dimensions: H1916 x W1494 x D960mm..Weight: 750kg..FIVE YEAR WARRANTY HARDWARE, 1 YEAR WARRA...