Learn More
Kuhner Shaker Inc SHAKER CABINET ISF4-XC WITHOUT
Catalog No. 502570051
Encompass
Shaker Cabinet ISF4-XC without cooling..Temperature range: ambient + 10C to 60C..Control of 0.1C, Accuracy +/- 0.3C at 37C..CO2 Control 0-20%. Control of 0.1%, Accuracy +/- 0.42% @ 20% CO2..100% stainless steel interior..External Dimensions: H1916 x W1...
Catalog No. 50-257-0051 Supplier Kuhner Shaker Inc Supplier No. SMX1600C
Add to Cart
Add to Cart
Request a Quote
Shaker Cabinet ISF4-XC without cooling..Temperature range: ambient + 10C to 60C..Control of 0.1C, Accuracy +/- 0.3C at 37C..CO2 Control 0-20%. Control of 0.1%, Accuracy +/- 0.42% @ 20% CO2..100% stainless steel interior..External Dimensions: H1916 x W1...