Learn More
Thomas Scientific VorTemp 56 Shaking Incubator, 115V
SDP
Catalog No. 501461146
Encompass
Designed for simultaneous heating and mixing of small samples, the VorTemp™ 56 features interchangeable platforms for microtubes and microplates. Both platforms are supplied with the unit. Temperature in the VorTemp™ 56 microplate shaking incubator is adjustable over a broad range, making the unit useful for a variety of applications, including bacterial cultures in tubes and microplates. Mechanical convection provides a uniform and stable environment within the chamber. The VorTemp™ 56 may also be used in the cold room for sub-ambient applications. Shake and incubate samples simultaneously Complete with platforms for microtubes and microplates Intelligent control Broad temperature and speed range Shaking speed as well as acceleration rates are adjustable. Acceleration can be set to three different levels to protect delicate samples. A microprocessor controls all parameters including temperature and timed operation. Self diagnostic software continuously monitors the u
Catalog No. 50-146-1146 Supplier Thomas Scientific Supplier No. 1197M96
Designed for simultaneous heating and mixing of small samples, the VorTemp™ 56 features interchangeable platforms for microtubes and microplates. Both platforms are supplied with the unit. Temperature in the VorTemp™ 56 microplate shaking incubator is adjustable over a broad range, making the unit useful for a variety of applications, including bacterial cultures in tubes and microplates. Mechanical convection provides a uniform and stable environment within the chamber. The VorTemp™ 56 may also be used in the cold room for sub-ambient applications. Shake and incubate samples simultaneously Complete with platforms for microtubes and microplates Intelligent control Broad temperature and speed range Shaking speed as well as acceleration rates are adjustable. Acceleration can be set to three different levels to protect delicate samples. A microprocessor controls all parameters including temperature and timed operation. Self diagnostic software continuously monitors the u