Learn More
Chemglass Life Sciences MultiTherm Heating and Cooling Shaker, 120V
SDP
Catalog No. 50143349
Encompass
MultiTherm Heating and Cooling Shaker, 120V. The Multi-Therm Shaker is a temperature controlled vortexer, useful for a variety of molecular biology applications including:¿ Denaturation of DNA, RNA and Proteins ¿ Reverse Transcription ¿ Lipid Extractions ¿ DNA/Plasmid Isolation ¿ Yeast and Bacteria Cultivation ¿ Gentle Thawing of Biological Materials ¿ Restriction DigestsDespite its modest 8 x 11¿ footprint, this Benchmark vortexer is compatible with all popular tubes and plates, including as many as six 50mL tubes. Speed, time and temperature settings are continuously visible on the LCD, simultaneously showing both actual and selected values. Integral over-temperature control ensures long life, safety and sample integrity
Catalog No. 50-143-349 Supplier Chemglass Life Sciences Supplier No. CLS4030001
MultiTherm Heating and Cooling Shaker, 120V. The Multi-Therm Shaker is a temperature controlled vortexer, useful for a variety of molecular biology applications including:¿ Denaturation of DNA, RNA and Proteins ¿ Reverse Transcription ¿ Lipid Extractions ¿ DNA/Plasmid Isolation ¿ Yeast and Bacteria Cultivation ¿ Gentle Thawing of Biological Materials ¿ Restriction DigestsDespite its modest 8 x 11¿ footprint, this Benchmark vortexer is compatible with all popular tubes and plates, including as many as six 50mL tubes. Speed, time and temperature settings are continuously visible on the LCD, simultaneously showing both actual and selected values. Integral over-temperature control ensures long life, safety and sample integrity