Learn More
Chemglass Life Sciences Orbital Shaker, 120V, Versa-Orb, Base Only, Digital
SDP
Catalog No. 50143325
Encompass
Multipurpose benchtop shaker.Accepts peg-and-grommet trays – sold separately.10 to 350rpm range with digital speed display.One-inch orbit.Two-year warranty.Built for versatility and performance, this multipurpose shaker base features a space-saving, low-profile design and is ideal for the agitation of gels, staining, de-staining and assay development.The peg-and-grommet-type top plate accepts a variety of easily installed trays and is compatible with similar trays on the market.Speed, controlled by a rotary knob with convenient locking ring, is adjustable from 10 to 350rpm and is indicated in a bright, digital display.The shaker boasts a 1-inch orbit and a maximum load of 25lbs.The 24V DC brushless motor is low-maintenance and features a detachable power supply.Two-year warranty included.Trays with liners are sold separately.Please Note: Flasks and Tray must be purchased separately.NOTE: Trays are not supplied with base and must be purchased separately.
Catalog No. 50-143-325 Supplier Chemglass Life Sciences Supplier No. CLS4021100
Multipurpose benchtop shaker.Accepts peg-and-grommet trays – sold separately.10 to 350rpm range with digital speed display.One-inch orbit.Two-year warranty.Built for versatility and performance, this multipurpose shaker base features a space-saving, low-profile design and is ideal for the agitation of gels, staining, de-staining and assay development.The peg-and-grommet-type top plate accepts a variety of easily installed trays and is compatible with similar trays on the market.Speed, controlled by a rotary knob with convenient locking ring, is adjustable from 10 to 350rpm and is indicated in a bright, digital display.The shaker boasts a 1-inch orbit and a maximum load of 25lbs.The 24V DC brushless motor is low-maintenance and features a detachable power supply.Two-year warranty included.Trays with liners are sold separately.Please Note: Flasks and Tray must be purchased separately.NOTE: Trays are not supplied with base and must be purchased separately.