Learn More
Grainger Benchmark Scientific Incu-Shaker 10L Shaking Incubator, 115V, 30 to 300 RPM, 1 ea
Catalog No. 502749816
Encompass_Preferred
The Incu-Shaker 10L is an orbital shaking incubator designed for reliable temperature control and consistent sample agitation. It features a robust design with two platforms, offering a versatile solution for applications requiring microplates or petri dishes.
  • 19 mm orbital movement
  • 30 to 300 RPM speed range
  • 5°C to 60°C operating temperature range
  • 120 V, 7.5 A, 900 W power requirements
  • 30 lb. (13.6 kg) weight capacity
  • Includes a non-slip rubber mat
  • CE compliant
Catalog No. 50-274-9816 Supplier Grainger Supplier No. 14N131
Add to Cart
Add to Cart
Request a Quote
The Incu-Shaker 10L is an orbital shaking incubator designed for reliable temperature control and consistent sample agitation. It features a robust design with two platforms, offering a versatile solution for applications requiring microplates or petri dishes.
  • 19 mm orbital movement
  • 30 to 300 RPM speed range
  • 5°C to 60°C operating temperature range
  • 120 V, 7.5 A, 900 W power requirements
  • 30 lb. (13.6 kg) weight capacity
  • Includes a non-slip rubber mat
  • CE compliant