Learn More
Grainger Benchmark Scientific Orbital Shaker, 14 lb Capacity, 30 to 300 RPM, 13 x 12 in. Platform
Catalog No. 502751752
Encompass_Preferred
This orbital shaker is designed for versatile laboratory mixing and aeration applications. It features a touch screen display and comes with a 33 cm x 30 cm (13 in. x 12 in.) non-slip, rubber coated platform ideal for tissue culture flasks, petri dishes, and staining trays. With a 19 mm orbital rotation and a load capacity of up to 6.4 kg (14 lb.), it provides reliable performance across a wide speed range.
  • Speed range of 30 rpm to 300 rpm with 1 rpm increments
  • Operating temperature range of +4°C to +65°C, suitable for cold room use
  • Programmable timer from 1 min. to 99 hr. or continuous operation
  • Includes a standard non-slip rubber mat platform
  • Compatible with optional magnetic attachment platform for Erlenmeyer flasks and test tube racks
  • CE compliant design
Catalog No. 50-275-1752 Supplier Grainger Supplier No. 14N122
This orbital shaker is designed for versatile laboratory mixing and aeration applications. It features a touch screen display and comes with a 33 cm x 30 cm (13 in. x 12 in.) non-slip, rubber coated platform ideal for tissue culture flasks, petri dishes, and staining trays. With a 19 mm orbital rotation and a load capacity of up to 6.4 kg (14 lb.), it provides reliable performance across a wide speed range.
  • Speed range of 30 rpm to 300 rpm with 1 rpm increments
  • Operating temperature range of +4°C to +65°C, suitable for cold room use
  • Programmable timer from 1 min. to 99 hr. or continuous operation
  • Includes a standard non-slip rubber mat platform
  • Compatible with optional magnetic attachment platform for Erlenmeyer flasks and test tube racks
  • CE compliant design