Learn More
Grainger Shaker Orbital 50-300 rpm 5 x10.75 x11
Catalog No. 502751750
Encompass_Preferred
The Benchmark Scientific Orbital Shaker is a compact and quiet orbital shaker designed for mixing small samples in the lab. It has a low profile design with rubber feet to prevent sliding and provide stability. The shaker has an adjustable speed from 30 to 300 rpm with a continuous or timed operation. The universal platform measures 14 x 14 inches and accommodates flasks up to 2 liters. This shaker is made from corrosion resistant materials for durability and includes both 115V and 230V motors for worldwide compatibility. With its small footprint, adjustable speed, and universal platform, this shaker provides reliable sample agitation for various lab applications.
Catalog No. 50-275-1750 Supplier Grainger Supplier No. 14N120
The Benchmark Scientific Orbital Shaker is a compact and quiet orbital shaker designed for mixing small samples in the lab. It has a low profile design with rubber feet to prevent sliding and provide stability. The shaker has an adjustable speed from 30 to 300 rpm with a continuous or timed operation. The universal platform measures 14 x 14 inches and accommodates flasks up to 2 liters. This shaker is made from corrosion resistant materials for durability and includes both 115V and 230V motors for worldwide compatibility. With its small footprint, adjustable speed, and universal platform, this shaker provides reliable sample agitation for various lab applications.