Learn More
Grainger Benchmark Scientific MultiTherm Shaker, Heating and Cooling, 200 to 1500 Rpm, Ambient -20°C to 100°C, 1 ea
Catalog No. 502749518
Encompass_Preferred
The MultiTherm shaker is a temperature-controlled vortexer designed for diverse molecular biology applications, including the denaturation of DNA, RNA, and proteins. This compact instrument features a precise heating and cooling system and is compatible with various tubes and plates ranging from 0.2 mL to 50 mL. An integrated LCD continuously displays both actual and selected speed, time, and temperature parameters for real-time monitoring.
  • Temperature range of ambient -20°C to 100°C with 0.1°C increments
  • Variable shaking speed of 200 to 1,500 rpm with a 3 mm orbit
  • Heating rate of 5.0°C/min. and cooling rate of 2.5°C/min.
  • Integral over-temperature control ensures safety and sample integrity
  • Programmable cycling with up to five steps and sequence link functions
  • Precision contoured wells for uniform thermal transfer
Catalog No. 50-274-9518 Supplier Grainger Supplier No. 14N136
The MultiTherm shaker is a temperature-controlled vortexer designed for diverse molecular biology applications, including the denaturation of DNA, RNA, and proteins. This compact instrument features a precise heating and cooling system and is compatible with various tubes and plates ranging from 0.2 mL to 50 mL. An integrated LCD continuously displays both actual and selected speed, time, and temperature parameters for real-time monitoring.
  • Temperature range of ambient -20°C to 100°C with 0.1°C increments
  • Variable shaking speed of 200 to 1,500 rpm with a 3 mm orbit
  • Heating rate of 5.0°C/min. and cooling rate of 2.5°C/min.
  • Integral over-temperature control ensures safety and sample integrity
  • Programmable cycling with up to five steps and sequence link functions
  • Precision contoured wells for uniform thermal transfer