Learn More
Grainger Shaker Vortex Mixer 200-3200rpm 6.7x6.3
Catalog No. 502752085
Encompass_Preferred
The Benchmark Scientific Analog Vortex Mixer is a compact and lightweight vortex mixer designed for continuous or touch operation. It features analog speed control with range up to 3000rpm and accommodates tubes up to 30mm diameter. The mixer has a stable metal base to prevent walking and rubber feet to reduce vibration. It requires 120V AC power and comes with a 3-prong plug for use in the US. The dimensions are 3.5"W x 5.5"D x 3.25"H. This analog vortex mixer provides reliable mixing performance for various lab applications.
Catalog No. 50-275-2085 Supplier Grainger Supplier No. 14N110
The Benchmark Scientific Analog Vortex Mixer is a compact and lightweight vortex mixer designed for continuous or touch operation. It features analog speed control with range up to 3000rpm and accommodates tubes up to 30mm diameter. The mixer has a stable metal base to prevent walking and rubber feet to reduce vibration. It requires 120V AC power and comes with a 3-prong plug for use in the US. The dimensions are 3.5"W x 5.5"D x 3.25"H. This analog vortex mixer provides reliable mixing performance for various lab applications.