Learn More
Kuhner Shaker Inc SHAKERINCUBATOR ISF1-Z WITH HE
Catalog No. 502569936
Encompass
Beluga Basic ZCTemperature range: ambient -15C to 80CControl of 0.1C, Accuracy +/- 0.3C at 37CUser Selectable Orbit Diameter (12.5mm, 25mm, 50mm)Loads up to 25kg (+25kg possible if required)DC direct drive with single eccentric (no belt)100% stainless steel interiorFIVE YEAR WARRANTY HARDWARE,1 YEAR WARRANTY ON SENSORS
Catalog No. 50-256-9936 Supplier Kuhner Shaker Inc Supplier No. SMZ1500BC
Add to Cart
Add to Cart
Request a Quote
Beluga Basic ZCTemperature range: ambient -15C to 80CControl of 0.1C, Accuracy +/- 0.3C at 37CUser Selectable Orbit Diameter (12.5mm, 25mm, 50mm)Loads up to 25kg (+25kg possible if required)DC direct drive with single eccentric (no belt)100% stainless steel interiorFIVE YEAR WARRANTY HARDWARE,1 YEAR WARRANTY ON SENSORS