Learn More
Kuhner Shaker Inc SHAKING CABINET ISF4-X WITH HU
Catalog No. 502570048
Encompass
Shaking Cabinet ISF4-X with humidity..Temperature range: ambient - 15C to 80C..Bi-directional humidity control to 90%RH, accuracy +/-1%..Control of 0.1C, Accuracy +/- 0.3C at 37C..100% stainless steel interior..External Dimensions: H1916 x W1494 x D960...
Catalog No. 50-257-0048 Supplier Kuhner Shaker Inc Supplier No. SMX1603
Add to Cart
Add to Cart
Request a Quote
Shaking Cabinet ISF4-X with humidity..Temperature range: ambient - 15C to 80C..Bi-directional humidity control to 90%RH, accuracy +/-1%..Control of 0.1C, Accuracy +/- 0.3C at 37C..100% stainless steel interior..External Dimensions: H1916 x W1494 x D960...