Learn More
MTC Bio Shaking incubator with CO2
Catalog No. NC1827681
Encompass_Preferred
Shaking Incubator, CO2, 30-300rpm, 19mm oribt, 0-20% CO2, Ambient+5 to 60ºC 46L, stainless steel interior with rounded corners, heated door with 6 sided heating, dual beam IR sensor, ideal for suspension cell culture
Catalog No. NC1827681 Supplier MTC Bio Supplier No. TB3501
Add to Cart
Add to Cart
Request a Quote
Shaking Incubator, CO2, 30-300rpm, 19mm oribt, 0-20% CO2, Ambient+5 to 60ºC 46L, stainless steel interior with rounded corners, heated door with 6 sided heating, dual beam IR sensor, ideal for suspension cell culture